Check Order Status
Astound ™ Fabric Reinforced Gowns
Actual product may vary
Astound ™ Fabric Reinforced Gowns
Protection, comfort and convenience
Have Questions?
Catalog number
9541
Price (MYR)
3,462.00
CS
Add to cart
Price (MYR)
3,462.00
CS
Add to cart
AAMI Level 3
Fabirc reinforcement in the sleeve and chest areas
Set-in sleeves
Sterile back
All sterile gowns include one hand towel per gown
Color: blue
Specifications
Gender
Unisex
No. of Pockets
None
Product Type
General Purpose Gown
Size
X-Large
Color
Blue
Disposable
Single-use
Sterility
Non-sterile
Autoclavable
Not Autoclavable