Search Thermo Fisher Scientific
Catalog Number | Color | Dispensing Speed |
---|---|---|
9521 | Blue | 8 |
9531 also known as BK953X1 | Red | 8 |
Carry out longer, fatigue-free pipetting with the Thermo Scientific™ S1 Pipette Fillers. These instruments are ergonomically designed and offer simple, efficient pipetting performance, as well as maximum pipetting comfort. Lightweight and cordless, they feature a heavy-duty rechargeable lithium-ion battery that allows approximately three times longer operation between charges than typical units. For use with graduated and volumetric glass and plastic serological pipets.
Monitor Performance
Control Speed
Ready When You Need It