ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
Actual product may vary
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip Style
94420153125 μLStandard 384
9442005312.5 μLStandard 384
9442010330 μLStandard 384
Catalog number 94420153
Price (MYR)
4,884.00
PK
Add to cart
Volume (Metric):
125 μL
Tip Style:
Standard 384
Price (MYR)
4,884.00
PK
Add to cart
Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovativesnap and seal tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.
Key Features:
  • All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
  • Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
  • Ideal for 384-format microplate applications
  • Filtered tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables
Immediate traceability
  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionClipTip 384 125, Filtered, Sterile, Rack
Volume (Metric)125 μL
FilteredFiltered
SterilitySterile
AutoclavableNot Autoclavable
Length (English)2.52 in.
Length (Metric)6.4cm
FormatRack
Tip StyleStandard 384
Packaging10 x 384 tips/rack
Color CodeYellow
For Use With (Equipment)384-Format Microplate, E1-ClipTip Electronic
Product LineClipTip
Fit for PurposeSterile & PCR Ready
Unit SizePK