Non-Filtered Extended Length Pipette Tips
Non-Filtered Extended Length Pipette Tips
Actual product may vary
Non-Filtered Extended Length Pipette Tips
Ensure a better seal with the pipetter module, for higher accuracy and precision.
Have Questions?
Catalog NumberVolume (Metric)Sterility
9406097010 mLNon-sterile
Catalog number 94060970
Price (MYR)
377.00
PK
Add to cart
Volume (Metric):
10 mL
Sterility:
Non-sterile
Price (MYR)
377.00
PK
Add to cart
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI).
Benefits
  • Excellent for single and multichannel pipetting
  • Available in six volume ranges: 0.2-10 μL, 1-200 μL, 5-300 μL, 100-1000 μL, 50-1200 μL and 10mL
  • Sterile tips are supplied in vacuum sealed, sterilized, 10 x 96 rack and they are certified to be free from DNA, DNase, RNase and Endotoxins
  • Save space with refills
Softer Tip
  • The soft tip reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Specifications
DescriptionFinntip Flex 10mL Ext, Non-Filtered, Non-sterile, Bulk
Volume (Metric)10 mL
FilteredNon-filtered
SterilityNon-sterile
ModelFinntip Flex 10mL Ext
Length (English)10.5 in.
Length (Metric)26.7cm
FormatBulk
Color CodeRed
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip Flex
Fit for PurposeCertified Quality
Unit SizePK