Finntip™ Wide Orifice Pipette Tips
Finntip™ Wide Orifice Pipette Tips
Actual product may vary
Finntip™ Wide Orifice Pipette Tips
Finntip™ Wide Orifice Pipette Tips
Finntip™ Wide Orifice Pipette Tips
Finntip™ Wide Orifice Pipette Tips
Eliminate cell fragmentation caused by mechanical shearing with these wide orifice pipette tips, ideal for pipetting macromolecules.
Have Questions?
Catalog NumberVolume (Metric)SterilityFormat
9405120250 μLNon-sterileRack
Catalog number 9405120
Price (MYR)
738.00
PK
Add to cart
Volume (Metric):
250 μL
Sterility:
Non-sterile
Format:
Rack
Price (MYR)
738.00
PK
Add to cart
Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips, ideal for pipetting macromolecules.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked or bulk
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Tip Range
  • Wide orifice tips available for volumes 250 and 1000 μL allowing better liquid handling for viscous liquids and sensitive cell samples
  • Inside diameter of orifice is 1.1mm/1.8mm
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 250 Wide, Non-Filtered, Non-sterile, Rack
Volume (Metric)250 μL
FilteredNon-filtered
SterilityNon-sterile
ModelFinntip 250 Wide
Length (English)2 in.
Length (Metric)5.3 cm
FormatRack
Tip StyleWide Orifice
Color CodeWhite
For Use With (Equipment)Thermo Scientific Finnpipette 250
Product LineFinnTip Wide Orifice
Fit for PurposeCertified Quality
Unit SizePK