Check Order Status
Accessories for Digital Mini Rotators
Actual product may vary
Accessories for Digital Mini Rotators
Get assorted accessories for use with Digital Mini Rotators
Have Questions?
Change view
Catalog Number
Description
Dimensions (L x W)
88882105
Universal (Aluminum) Platform
310 x 284 mm
88882110
1 L Flask Clamp
130 x 77 mm
88882104
Universal Attachment
307 x 303 mm
3 Options
Catalog number
88882105
Price (MYR)
807.00
EA
Add to cart
Description:
Universal (Aluminum) Platform
Dimensions (L x W):
310 x 284 mm
Price (MYR)
807.00
EA
Add to cart
Assorted accessories for use with Digital Mini Rotators
Specifications
Product Type
Universal Platform
Certifications/Compliance
Certificate of Analysis (COA), Certificate of Conformance (COC)
Material
Aluminum
Dimensions (L x W)
310 x 284 mm
Length (English) Platform
12.21 in.
Length (Metric) Platform
310 mm
Width (English) Platform
11.18 in.
Width (Metric) Platform
284 mm
Description
Universal (Aluminum) Platform
For Use With (Equipment)
Digital Mini Rotators, Rocker, Rotator, Mixer
Warranty and Service Offering
2 Years