Digital Microplate Shakers
Digital Microplate Shakers
Actual product may vary
Digital Microplate Shakers
Digital Microplate Shakers
Digital Microplate Shakers
Digital Microplate Shakers
Get accurate and efficient sample mixing
Have Questions?
Catalog NumberPlug TypeVoltage
88882006EU, UK, CN Plug230 V
Catalog number 88882006
Price (MYR)
5,549.00
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Voltage:
230 V
Price (MYR)
5,549.00
EA
Add to cart
Stable and accurate microplate shaking with standard and enlarged platform that can shake maximum of six microplates at one run.
  • Digital display of Speed and Time
  • Speed range: 150 – 1200 rpm
  • Precise speed control:
  • ±1% of set speed up to 299rpm
  • ±2% 300 to 1200rpm
  • Orbit Diameter: 2.5mm
  • Max load up to 4 kg
  • Timer mode: up to 99h 59min
  • Optional large platform applied for 6 microplates
  • Optional tube racks applied for 1.5ml, 15ml and 50ml tubes
Specifications
Orbit2.5 mm
Load Bearing Capacity (Metric)1 kg at 1200 rpm, 4 kg at ≤400 rpm
Speed Control Accuracy1 rpm
Speed Range150 to 1200 rpm
Electrical Requirements100/240 V
Voltage230 V
Temperature Range (Metric)5°C to 40°C
Amperage72 VA
Product TypeDigital Microplate Shaker
Certifications/CompliancecCSAus, CE, RoHS
Plug TypeEU, UK, CN Plug
Temperature Range (English)41°F to 104°F
Frequency50/60 Hz
MaterialStainless Steel Platform
Dimensions (D x W x H) Interior383 x 260 x 145 mm (15.1 x 10.2 x 5.7 in.)
Dimensions (LxW) Platform275 x 235 mm (10.8 x 9.3 in.)
Dimensions (L x W x H)487 x 387 x 264mm (19.2 x 15.2 x 10.4 in.)
Length (English) Platform10.8 in.
Length (Metric) Platform275 mm
Width (English) Platform9.3 in.
Width (Metric) Platform235 mm
Shipping Weight (English)33 lb.
Shipping Weight (Metric)15 kg / 33 lbs
Weight (English)28.2 lb.
Weight (Metric)12.8 kg
Controller TypeLED
ModelDigital Microplate Shaker
DisplayLED
For Use With (Equipment)Rocker, Rotator, mixer
Humidity Range20 to 80%
Platform StyleTray
Timer Range1 min. to 99:59 hr.
Unit SizeEA