Digital Rockers
Digital Rockers
Actual product may vary
Digital Rockers
Digital Rockers
Digital Rockers
Digital Rockers
Mix samples accurately and efficiently
Have Questions?
Catalog NumberProduct TypePlug TypeIncludesVoltage
88882002Digital RockerEU, UK, CN PlugDimpled Rubber Mat tray x 1, Rubber strip x 6, User Manual x 1, EU plug x 1, UK plug x 1, CHN plug x 1230 V
Catalog number 88882002
Price (MYR)
4,894.00
EA
Add to cart
Product Type:
Digital Rocker
Plug Type:
EU, UK, CN Plug
Includes:
Dimpled Rubber Mat tray x 1, Rubber strip x 6, User Manual x 1, EU plug x 1, UK plug x 1, CHN plug x 1
Voltage:
230 V
Price (MYR)
4,894.00
EA
Add to cart
Digital display with big platform, accurate speed control and quick angle adjustment for easy sample agitation
  • Digital display of Speed and Time
  • Speed range: 1 – 70 rpm
  • Easily adjustable rocking angle: 1°- 15°
  • Precise speed control:
  • ±1 rpm ≤ 50 rpm
  • ±2 rpm > 50rpm
  • Max load up to 4.5 kg
  • Tray size fits 6 x well-plates
  • Timer Range: Continuous or 1 sec to 9999 min
  • Optional Non-slip rubber mat tray is designed for carrying plates, dishes, culture bottles and bags
Specifications
Rocking Angle1 to 15°
Load Bearing Capacity (Metric)4.5 kg
Speed Control Accuracy±1 rpm (≤ 50 rpm), ±2 rpm (>50 rpm)
Speed Range1 to 70 rpm
Electrical Requirements100/240 V
Voltage230 V
Amperage0.1 A
Product TypeDigital Rocker
Load Bearing Capacity (English)9.92 lb.
Certifications/CompliancecCSAus, CE, RoHS
Plug TypeEU, UK, CN Plug
Temperature ControlDigital
Frequency50/60 Hz
MaterialStainless Steel Platform
Dimensions (D x W x H) Interior390 x 292 x 206 mm (15.4 x 11.5 x 8.1 in.)
Dimensions (LxW) Platform322 x 282 mm (12.7 x 11.1 in.)
Dimensions (L x W x H)487 x 387 x 354 mm (19.2 x 15.2 x 13.9 in.)
Height (English)13.9 in.
Height (Metric)354 mm
Shipping Weight (English)29.8 lb.
Shipping Weight (Metric)13.5 kg
Controller TypeLED
ModelDigital Rocker
DisplayLED
Humidity Range20 to 85%
IncludesDimpled Rubber Mat tray x 1, Rubber strip x 6, User Manual x 1, EU plug x 1, UK plug x 1, CHN plug x 1
Platform StyleTray
Timer Range1 sec. to 9999 min., Continuous
Warranty and Service Offering2 Years
Unit SizeEA