Clamps for CO2 Resistant Shaker
Clamps for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClampsProduct Type
88881133FlaskStarter Kit
88881107150 mLFlask Clamp
88881109250 mLFlask Clamp
88881110500 mLFlask Clamp
888811111000 mL Flask Clamp
888811122000 mLFlask Clamp
Catalog number 88881133
Price (MYR)
2,208.00
EA
Add to cart
Clamps:
Flask
Product Type:
Starter Kit
Price (MYR)
2,208.00
EA
Add to cart
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Product TypeStarter Kit
ClampsFlask
For Use With (Equipment)CO2 Resistant Shaker
Includes(2) 100mL flask clamps, (4) 250mL flask clamps, (4) 500mL flask clamps, (2) 1L flask clamps, (2) 2L flask clamps
No. of PositionsVaries
Unit SizeEA