Racks and Accessories for CO2 Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Racks and Accessories for CO2 Resistant Shaker
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
88881132UK Plug
88881119Fixed Tube Rack 40 x 16 mm dia.
88881122Universal aluminum platform
88881123Rubber mat platform
8888112496-Well microplate holder
88881126Sticky mat 140 x 140 mm
Catalog number 88881132
Price (MYR)
57.00
EA
Add to cart
Includes:
UK Plug
Price (MYR)
57.00
EA
Add to cart
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Plug TypeUK Plug
For Use With (Equipment)CO2 Resistant Shaker
IncludesUK Plug
Unit SizeEA