Accessories for Digital Heating Shaking Baths
Accessories for Digital Heating Shaking Baths
Actual product may vary
Accessories for Digital Heating Shaking Baths
Use Thermo Scientific Accessories for Digital Shaking Baths to optimize flexibility and performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of PlacesTube Size
88880128650mL
88880125960.2mL
88880126480.5mL
88880127241.5mL
Catalog number 88880128
Price (MYR)
2,674.00
EA
Add to cart
No. of Places:
6
Tube Size:
50mL
Price (MYR)
2,674.00
EA
Add to cart
Use Thermo Scientific Accessories for Digital Heating Shaking Baths to optimize flexibility and performance.
  • Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (Metric)50 mL
For Use With (Equipment)Heating/Cooling Bath
LightingLED
No. of Places6
TypeBlock Set with Cover
Tube Size50mL
Unit SizeEA