Digital Heating Cooling Drybath
Digital Heating Cooling Drybath
Actual product may vary
Digital Heating Cooling Drybath
Maximize lab space and flexiblity with our compact, programmable heating/cooling bath with digital display.
Have Questions?
Catalog NumberElectrical RequirementsPlug Type
88880030230 V, 50/60 HzCord set w/various plugs
Catalog number 88880030
Price (MYR)
9,030.00
EA
Add to cart
Electrical Requirements:
230 V, 50/60 Hz
Plug Type:
Cord set w/various plugs
Price (MYR)
9,030.00
EA
Add to cart
Maximize lab space and flexiblity with the compact, programmable Thermo Scientific™ Digital Heating Cooling Drybath. PID control ensures consistent precise temperature control. Drybath has a low-profile design and two types of timing modes.
PID control ensures consistent precise temperature control.
  • Temperature Range: 4° to 95°C
  • Program Storage:
    • 10 programs, up to 10 steps per program
  • Two types of timing modes:
    • Start only after temperature reaches desired set point
    • Start immediately after the timer is set
  • Low-profile design
  • Blocks easily interchanged
Ordering Information:
  • Drybath requires blocks; must be purchased separately
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE
Controller TypeFeedback Control PID
LightingLED
Plug TypeCord set w/various plugs
Temperature Range (Metric)4°C to 95°C
TypeDry Bath Block
DisplayDigital
Temperature Accuracy0.55°C at 37°C
Temperature Stability±0.15 at 37°C
Temperature Uniformity±0.55 at 37°C
Warranty2 Years
Dimensions (L x W x H)13.1 x 10.9 x 6.5 in. (334 x 276 x 165 mm)
Dimensions (LxWxH) Interior3.1 x 3.9 x 1.4 in. (77.5 x 99 x 36 mm) (Interior)
Shipping Weight (English)11 lb.
Shipping Weight (Metric)5 kg
Electrical Requirements230 V, 50/60 Hz