Digital Heating Shaking Drybath
Digital Heating Shaking Drybath
Actual product may vary
Digital Heating Shaking Drybath
Increase lab versatility with our digital heating shaking drybath, which can be used as a drybath, shaker or both.
Have Questions?
Catalog NumberElectrical Requirements
88880028230 V, 50/60 Hz
Catalog number 88880028
Price (MYR)
8,021.00
EA
Add to cart
Electrical Requirements:
230 V, 50/60 Hz
Price (MYR)
8,021.00
EA
Add to cart
Increase lab versatility with the Thermo Scientific™ Digital Heating Shaking Drybath, which can be used as a drybath, shaker or both. PID control ensures consistent shaking results and precise temperature control. Drybath has a low-profile design and two types of timing modes.
PID control ensures consistent shaking results and precise temperature control.
  • Speed range from 150 to 1500rpm (block dependent)
  • Program Storage:
    • 10 programs, up to 10 steps per program
    • Interval shaking function allows shaking and nonshaking in same program
  • Two types of timing modes:
    • Start only after temperature reaches desired set point
    • Start immediately after the timer is set
  • Low-profile design
  • Easily changed blocks in multiple sizes for maximum flexibility
Ordering Information:
  • Drybath requires blocks; must be purchased separately
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE
Controller TypeFeedback Control PID
Capacity (Metric)0.5, 1.5, 50 mL
LightingLED
No. of Places96
Plug TypeCord set w/various plugs
Temperature Range (Metric)Ambient +5° to 100°C
TypePlatform Shaker
DisplayDigital
No. of BlocksNot included; purchase separately
Speed150 to 1,500rpm (96-well block); 150 to 1,200rpm (0.5mL, 1.5mL and 50mL tube blocks)
Temperature Accuracy0.5°C (0.9°F) at 80°C
Warranty2 Years
Dimensions (L x W x H)13.1 x 10.9 x 6.7 in. (334 x 276 x 170 mm)
Diameter (Metric) Orbit2 mm
Shipping Weight (English)18.3 lb.
Shipping Weight (Metric)8.3 kg
Electrical Requirements230 V, 50/60 Hz