RT2 Hotplates, Basic Model
RT2 Hotplates, Basic Model
Actual product may vary
RT2 Hotplates, Basic Model
Increase your confidence in safety and accuracy with our low-profile digital display hotplate.
Have Questions?
Catalog NumberTop Plate Material
88880001Aluminum, Ceramic
Catalog number 88880001
Price (MYR)
3,456.00
EA
Add to cart
Top Plate Material:
Aluminum, Ceramic
Price (MYR)
3,456.00
EA
Add to cart
Increase your confidence in safety and accuracy with the Thermo Scientific™ RT2 Hotplate, which offers a low-profile and digital display. It is equipped with hot top warning and built-in temperature shut-off to provide accurate heating of samples and safety to the laboratory.
In addition to a hot top warning indicator, an overheat prevention circuit turns off the heater if the top plate temperature reaches 450°C.
  • Digital control
  • Microprocessor PID temperature control
  • Three user-selectable temperature control modes:
    • Optimal Mode
    • Fast Mode
    • Slow Mode
  • Two-user selective timer to display actual and set value simultaneously
  • Nonslip heating bath (optional)
  • Transparent safety shield (optional)
Specifications
Body MaterialCast Aluminum
Top Plate MaterialAluminum, Ceramic
Temperature (English)122°F to 662°F
Controller TypePID
Size5.5 in. dia.
Temperature (Metric)50°C to 350°C
Height (English)3.94 in.
Length (English)11.42 in.
Width (English)6.34 in.
Height (Metric)100 mm
Length (Metric)290 mm
Width (Metric)161 mm
Wattage600 w
Electrical Requirements230 V, 50/60 Hz
Frequency50/60 Hz
Plug TypeVarious Styles
Max. Sample Load25 kg
ShapeRound
Voltage230 V
TypeHot Plate
Certifications/CompliancecCSAus, CE
Warranty2 Years
DisplayLED Digital
Dimensions (L x W x H)11.42 x 6.34 x 3.94 in. (290 x 161 x 100 mm)
Unit SizeEA