96-well/Flat Bottom Block
96-well/Flat Bottom Block
Actual product may vary
96-well/Flat Bottom Block
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Have Questions?
Catalog NumberNo. of Places
8887110796
Catalog number 88871107
Price (MYR)
822.00
EA
Add to cart
No. of Places:
96
Price (MYR)
822.00
EA
Add to cart
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
Specifications
Description96-well/flat bottom plate block
Capacity (Metric)0.2 mL
For Use With (Equipment)Thermo Scientific™ Compact Dry Bath/Block Heater
LightingLED
No. of Places96
TypeBlock
Dimensions (L x W x H)108 x 72 x 18 mm (4.3 x 2.8 x 0.7 in.)
Depth (Metric)13 mm
Unit SizeEA