Block, 6 x 15 mL
Product Image
Block, 6 x 15 mL
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Have Questions?
Catalog NumberNo. of Places
888711056
Catalog number 88871105
Price (MYR)
661.00
EA
Add to cart
No. of Places:
6
Price (MYR)
661.00
EA
Add to cart
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
Specifications
Description6 x 15mL block
Capacity (Metric)15 mL
For Use With (Equipment)Thermo Scientific™ Compact Dry Bath/Block Heater
LightingLED
No. of Places6
TypeDry Block
Dimensions (L x W x H)36 x 108 x 72 mm (1.4 x 4.3 x 2.8 in.)
Unit SizeEA