96-well Half/Full-Skirted PCR Plate Block
96-well Half/Full-Skirted PCR Plate Block
Actual product may vary
96-well Half/Full-Skirted PCR Plate Block
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog number 88870121
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
Description96-well half / full-skirted PCR
Capacity (Metric)0.2 mL
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds1 x 96 well half or full-skirted PCR plate
LightingLED
No. of Places96
TypeBlock
Dimensions (L x W x H)76 x 124 x 55 mm (3 x 4.9 x 2.17 in.)
Unit SizeEA