Polyester Adhesive Warning Sign: STORAGE AREA FLAMMABLE MATERIAL
Polyester Adhesive Warning Sign: STORAGE AREA FLAMMABLE MATERIAL
Actual product may vary
Polyester Adhesive Warning Sign: STORAGE AREA FLAMMABLE MATERIAL
Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
Have Questions?
Catalog NumberDimensions (L x W)
852-0014 x 10 in.
Catalog number 852-00
Price (MYR)
-
Dimensions (L x W):
14 x 10 in.
  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications
FinishLaminated
MaterialPolyester
Quantity1 Ea.
Adhesive TypeCold-temperature Permanent Acrylic
Mounting TypeSelf-adhesive
Height (English)10 in.
Max. Temperature (English)230°F
Min. Temperature (English)-40°F
Thickness0.01 in.
Width (English)14 in.
ShapeRectangular
Temperature Range (English)-40°F to 230°F
Dimensions (L x W)14 x 10 in.
Displayed TextSTORAGE AREA FLAMMABLE MATERIAL
ColorRed on White
Unit SizeEach