Validation Tools for UV-Visible Spectrophotometers
Validation Tools for UV-Visible Spectrophotometers
Actual product may vary
Validation Tools for UV-Visible Spectrophotometers
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Have Questions?
Catalog NumberType
840-326100Performance Verification Kit
Catalog number 840-326100
Price (MYR)
18,684.00
EA
Add to cart
Type:
Performance Verification Kit
Price (MYR)
18,684.00
EA
Add to cart
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Packages provide:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance
  • Installation and instrument certification
Specifications
TypePerformance Verification Kit
For Use With (Equipment)GENESYS 40-180, BioMate 160 Spectrophotometers
Unit SizeEA