VISIONlite™ Spectrophotometer Software
VISIONlite™ Spectrophotometer Software
Actual product may vary
VISIONlite™ Spectrophotometer Software
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Have Questions?
Catalog NumberType
840-312300VISIONlite 5 Software + Cable
Catalog number 840-312300
Price (MYR)
8,176.00
EA
Add to cart
Type:
VISIONlite 5 Software + Cable
Price (MYR)
8,176.00
EA
Add to cart
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite Software enhancements include:
Scan:
  • Peak pick to identify and label wavelength peaks or valleys
  • Import, save or export data in native or CSV format for easy archiving and reporting
  • Auto scaling of data plus simple zoom controls for detailed examination of spectra 
Rate:  For SPECTRONIC 200 and GENESYS 30, measure multi-cell kinetics in parallel or serial mode
  • Automatic enzyme activity calculation
  • Automatic data storage plus import or export your data in CSV format
  • Measure up to 100 samples in a single data set 
Fixed:
  • Measure at one or multiple wavelengths
  • Apply factors and equations to your data to generate results directly
  • Addition, subtraction, ratio and baseline correction options
  • Run standard bio-tests using the preprogrammed protein and DNA methods
Quant: More standards and non-linear curve fitting options than can be accomplished under local control on Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers
Specifications
TypeVISIONlite 5 Software + Cable
For Use With (Equipment)Thermo Scientific GENESYS 40-180 Spectrophotometers, SPECTRONIC 200 and GENESYS 30
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean
Required AccessoriesInterface cable included for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers
Unit SizeEA