Thermostatted Single Cell Holder for GENESYS 140/150/180 & BioMate 160 Spectrophotometers
Actual product may vary
Thermostatted Single Cell Holder for GENESYS 140/150/180 & BioMate 160 Spectrophotometers
Control sampling temperatures for life science applications such as biochemical kinetics, enzymatic food analysis and equilibrium studies.
Have Questions?
Catalog Number
For Use With (Equipment)
Includes
840-306800
GENESYS 140/150/180,BioMate160
1-Cell Holder + UK Power Cable
Catalog number 840-306800
Price (MYR)
18,176.00
EA
Add to cart
For Use With (Equipment):
GENESYS 140/150/180,BioMate160
Includes:
1-Cell Holder + UK Power Cable
Price (MYR)
18,176.00
EA
Add to cart
Ideal for life science applications such as biochemical kinetics, enzymatic food analysis and equilibrium studies.
Air-cooled Peltier thermostatted single cell holder for GENESYS 140/150/180 and BioMate 160 Spectrophotometers. The accessory fits inside the sample compartment of the instrument.
Includes cell holder for 10 mm pathlength square cells
Temperature range 20° C to 60° C
Fully controlled by the instrument on-board software or Thermo Scientific™ VISIONlite™ Software
Magnetic stirring (variable speed) for full size cells
Suitable for use with unmasked cells >= 1000 μL nominal volume
Microcells < 1000 μL must be masked type
Additional 10 pack of PTFE-coated cuvette stir bars (5 mm x 2 mm) available
Multi-voltage power supply
Specifications
TypeThermostatted Peltier Single Cell Holder
For Use With (Equipment)GENESYS 140/150/180,BioMate160