Check Order Status
GENESYS Fiber Optic Probes
Actual product may vary
GENESYS Fiber Optic Probes
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Have Questions?
Catalog Number
For Use With (Equipment)
Type
840-305700
GENESYS 150/180 Spectrophotometer
Fiber Optic Dip Probe System
Catalog number
840-305700
Price (MYR)
35,717.00
EA
Add to cart
For Use With (Equipment):
GENESYS 150/180 Spectrophotometer
Type:
Fiber Optic Dip Probe System
Price (MYR)
35,717.00
EA
Add to cart
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.
Specifications
Diameter (Metric)
6 mm
Type
Fiber Optic Dip Probe System
For Use With (Equipment)
GENESYS 150/180 Spectrophotometer