BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
Actual product may vary
BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
BioMate™ 160 UV-Vis Spectrophotometer
Perform automated measurements of nucleic acids, proteins and cell cultures with the Thermo Scientific BioMate 160 UV-Vis spectrophotometer.
Have Questions?
Catalog NumberTypeWarranty
840-301400UV-Vis Spec + UK Power Cable2 yrs std + 1 yr ext upon reg
Catalog number 840-301400
Price (MYR)
62,134.00
EA
Add to cart
Type:
UV-Vis Spec + UK Power Cable
Warranty:
2 yrs std + 1 yr ext upon reg
Price (MYR)
62,134.00
EA
Add to cart
Perform quantitative measurements in life science laboratories with the automated and network-capable Thermo Scientific™ BioMate™ 160 UV-Visible Spectrophotometer. The BioMate 160 Spectrophotometer offers pre-programmed assay methods for nucleic acid and protein concentration as well as cell culture measurements. Optimized for usability and performance, this platform features a high-resolution color touchscreen, optional Wi-Fi networking, and a rugged design.
The BioMate UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience. Software includes pre-programmed life science methods such as Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays and OD600.
A full line of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control.
Features:
  • 7-inch high-resolution touchscreen user interface
  • Pre-programmed life science methods
  • Networking capable (Wi-Fi adaptor available)
  • Room light resistance for 'open-lid' operation
  • Sipper accessory
  • Removable, washable sample compartment liner with magnetic placement and hold-down
  • Accommodates cells up to 100 mm pathlength
  • (Optional) 8-position cell changer with switchable turret
  • (Optional) 4-position cell changer (long path cells)
  • (Optional) Peltier-thermostatted single cell holder (20 °C - 60 °C)
Designed for:
  • Advanced teaching labs
  • R&D departments
  • Applications requiring high throughput or temperature control
Specifications
Baseline Flatness±0.002 A
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
ConnectionsSingle USB-A
Duplex USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7 in. High Definition Color Touchscreen, Tiltable, 800 x 1280 pixels
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Spectral Bandwidth2 nm
TypeUV-Vis Spec + UK Power Cable
Wavelength Range190 to 1100 nm
Drift(At 500 nm after 1 hour warmup) <0.0005 A/hr.
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
KeypadTouchscreen
Lamp Life> 5 years typical, 3 years guaranteed
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual Beam
Photometric Accuracy±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A
Photometric Display–3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001 A at 1 A measured at 1.0 A at 546 nm
Printer IncludedSnap-on printer available
Scan SpeedSlow, medium and fast - up to 1600 nm/min.
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Warranty2 yrs std + 1 yr ext upon reg
Wavelength Accuracy±0.5 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability±0.2 nm
Weight (Metric)7.5 kg
Unit SizeEA