GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
Actual product may vary
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
840-278700Tungsten Halogen Lamp
840-278000Thermal Snap-on Printer
840-259800Self-Stick Printer Paper
840-278300Standard Printer Paper
840-278500Replacement Compartment Lid
Catalog number 840-278700
Price (MYR)
1,247.00
EA
Add to cart
Type:
Tungsten Halogen Lamp
Price (MYR)
1,247.00
EA
Add to cart
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible In addition to valuable built-in features such as the high resolution color display, large sample compartment, and data export options, a complete selection of accessories are available for the the GENESYS 30 spectrophotometer to help meet the needs of students, educators and lab technicians for accurate quantitative analysis.
  • Holds one rectangular longpath cell 20 - 100mm pathlength 
  • Comes assembled in a removable sample compartment liner
  • Allows for flexibility between samples
Specifications
TypeTungsten Halogen Lamp
For Use With (Equipment)GENESYS 30 Visible Spectrophotometer
Includes1 pre-aligned lamp assembly
Unit SizeEA