Check Order Status
Holmium and Didymium Glass Standards
Holmium and Didymium Glass Standards
Thermo Scientific™ Wavelength Calibration Filter Sets consist of Holmium and Didymium solid filters traceable to international standards.
Have Questions?
Catalog Number
For Use With (Equipment)
840-253200
Standard 10mm pathlength square cuvette holder in any spectrophotometer
Catalog number
840-253200
Price (MYR)
-
For Use With (Equipment):
Standard 10mm pathlength square cuvette holder in any spectrophotometer
Thermo Scientific™ Wavelength Calibration Filter Sets consist of Holmium and Didymium solid filters traceable to international standards.
Calibrated according to a certified process in our NVLAP accredited calibration laboratory in Madison, WI, USA. (NVLAP Code: 200955-0)
Mounted in a filter holder suitable for use with any UV-Visible spectrophotometer sample holder
Calibrated at eight peak wavelengths for Holmium and five for Didymium
Specifications
Certifications/Compliance
Traceable to mercury emission lines, a recognized fundamental standard
Description
Holmium Didymium Glass Standards
For Use With (Equipment)
Standard 10mm pathlength square cuvette holder in any spectrophotometer