Evolution™ Smart Thermostatted Linear 8-Cell Changer Accessory
Evolution™ Smart Thermostatted Linear 8-Cell Changer Accessory
Actual product may vary
Evolution™ Smart Thermostatted Linear 8-Cell Changer Accessory
Precisely position up to eight samples with the Thermo Scientific™ Evolution™ Smart Thermostatted Linear 8-Cell Changer Accessory
Have Questions?
Catalog NumberFor Use With (Equipment)Type
840-219000Evolution SpectrophotometersLinear 8-Cell Changer
Catalog number 840-219000
Price (MYR)
28,521.00
EA
Add to cart
For Use With (Equipment):
Evolution Spectrophotometers
Type:
Linear 8-Cell Changer
Price (MYR)
28,521.00
EA
Add to cart
Precisely position up to eight samples with the Thermo Scientific™ Evolution™ Smart Thermostatted Linear 8-Cell Changer Accessory for the Thermo Scientific™ Evolution™ spectrophotometers. Thermostatting, where required, is accomplished with a flow of water from an external temperature controlled recirculating water bath.
Make the most of your spectrophotometer’s capabilities:
  • Automate measurement of up to 8 samples
  • Automate measurement of standards for QA/QC measurements
  • Measure multi-cell kinetics reactions
Features:
  • Temperature range: -10° to 100°C
  • Precision cell holder mechanism securely holds up to 8 standard 10mm pathlength cells
  • Automatic recognition and alignment
  • Precision engineered cell transport system offers quiet operation and positioning accuracy
  • Easy connections for circulating liquid temperature control
  • Collection tray and drain catches any leaking samples
  • Full software control with Thermo Scientific™ INSIGHT™ software 
Configurations:
  • Use in sample position of Evolution One/One Plus or Evolution Pro only
Options:
  • Recirculator package for temperature control
  • Appropriate temperature probe(s) for temperature monitoring
NOTE: Microcell and semi-microcell applications with this accessory are not recommended
Specifications
CalibrationCustom calibration sequence in INSIGHT software for the Evolution One/One Plus spectrophotometers finds the center of each cell aperture and remembers the position, going to that spot precisely each time it measures the cell.
Compatibility10mm pathlength standard volume, <10mm pathlength cells with appropriate spacer
Configuration DetailsSample position only
TypeLinear 8-Cell Changer
For Use With (Equipment)Evolution Spectrophotometers
Unit SizeEA