Acclaim™ Trinity Q1 LC Columns
Acclaim™ Trinity Q1 LC Columns
Actual product may vary
Acclaim™ Trinity Q1 LC Columns
Use to separate the herbicides diquat and paraquat.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Particle Size
832442.1 mm10 mm5 μm
Catalog number 83244
Price (MYR)
-
Diameter (Metric):
2.1 mm
Length (Metric):
10 mm
Particle Size:
5 μm
Simultaneously separate the herbicides diquat and paraquat with Thermo ScientificTM AcclaimTM Trinity Q1 LC Columns. These tri-mode (WCX, WAX, RP) columns offer unmatched high-resolution and high-throughput trace analysis through LC-MS/MS and LC-UV methods. The stationary phase, based on Nanopolymer Silica Hybrid (NSH) technology, provides multiple retention mechanisms, including reversed-phase, anion-exchange, and cation-exchange, for maximum flexibility in method development. Easily optimize the selectivity to accelerate chromatographic separations and increase productivity.
Superior Performance for Diquat and Paraquat Analysis
  • Distinctive spatial separation of anion-exchange and cation-exchange regions for unparalleled chromatographic performance
  • Adjustable selectivity using mobile phase buffer concentration, pH, and solvent content
  • Quick separation of hydrophilic ionic and ionizable analytes without ion-pairing reagent
  • Ideal selectivity for simultaneous separation of basic, neutral, and acidic analytes
  • Compatibility with mass spectrometry detection, which significantly improves sensitivity and provides conformation at the same time
  • Available in 3.0μm particle size and 2.1 and 3.0mm diameters, in 50mm length
About the Family of Acclaim Specialty HPLC Columns
Acclaim Specialty Columns have been developed, manufactured and tested to meet the separation needs of specific applications. These columns are based on novel and unique chemistries and provide superior resolution with ease-of-use.
  • Use-tested to guarantee consistent separations
  • Ideal selectivity for separating specific target analytes
  • Excellent peak shapes
  • Performance tested to ensure high quality and reliability for specific chromatographic separations
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
QuantityPack of 2
pH2.5 to 7.5
Particle Size5 μm
Length (Metric)10 mm
Pore Size300Å
Surface Area100 m2/g
Max. Pressure6000 psi
Diameter (Metric)2.1 mm
Diameter (Metric) Inner2.1 mm
Temperature (Metric) Maximum40°C
Column FormatGuard Cartridge, 2 Pk.
Column TypeSpecialty / Application Specific
EndcappedYes
Product LineAcclaim
Unit SizeEach