Glass Lined Stainless-Steel GC Tubing
Glass Lined Stainless-Steel GC Tubing
Actual product may vary
Glass Lined Stainless-Steel GC Tubing
Glass Lined Stainless-Steel GC Tubing
Glass Lined Stainless-Steel GC Tubing
Glass Lined Stainless-Steel GC Tubing
Exhibits resistance to the harshest of conditions and to strong acids and bases
Have Questions?
Catalog NumberOverall Length (Metric)Diameter (Metric) Inner
8276860 cm4 mm
Catalog number 82768
Price (MYR)
-
Overall Length (Metric):
60 cm
Diameter (Metric) Inner:
4 mm
  • Ideal for HPLC columns, particularly for biological matrices
  • Temperature stable up to 500°C
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use WithHPLC Columns, Biological Matrices
Diameter (Metric) Inner4 mm
Overall Length (Metric)60 cm
MaterialBorosilicate Glass (Lining)
Max. Operating Temperature (Metric)Stable up to 500°C
Unit SizeEach