Check Order Status
Glass Lined Tubing
Glass Lined Tubing
Glass Lined Tubing (GLT) is borosilicate glass lined stainless steel tubing giving excellent resistance to the harshest of conditions
Have Questions?
Catalog Number
Overall Length (Metric)
Diameter (Metric) Inner
Diameter (Metric) Outer
82767
30 cm
4 mm
6.35 mm
Catalog number
82767
Price (MYR)
-
Overall Length (Metric):
30 cm
Diameter (Metric) Inner:
4 mm
Diameter (Metric) Outer:
6.35 mm
Excellent reistance to strong acids and bases
Ideal for HPLC columns - particularly for biological matrices
Temperature stable up to 500°C
Specifications
Type
Tubing
Diameter (English) Outer
1/4 in.
Diameter (Metric) Inner
4 mm
Diameter (Metric) Outer
6.35 mm
Overall Length (Metric)
30 cm
Material
Stainless Steel, Borosilicate Glass (Lining)
Max. Operating Temperature (Metric)
500°C