TSKgel™ Amide-80 HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ Amide-80 HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™
Actual product may vary
TSKgel™ Amide-80 HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™
For the analysis of saccharides, glycans, oligosaccarides, peptides, and polar compounds from natural products.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
82001010 cm1 mm
Catalog number 820010
Price (MYR)
187.00
EA
Add to cart
Length (Metric):
10 cm
Diameter (Metric):
1 mm
Price (MYR)
187.00
EA
Add to cart
  • Columns are packed with 3, 5, or 10 micron spherical silica particles covalently bonded with carbamoyl groups
  • Amide stationary phase provides a unique selectivity under regular normal phase conditions, or in the hydrophilic interaction mode of chromatography
  • Target applications include the analysis of saccharides, glycans, oligosaccarides, peptides, and polar compounds from natural products
  • Unique advantages of Amide-80 phase for saccharide analysis include a novel hydrogen bonding retention mechanism between hydroxyl groups of the sample and the carbamoyl group in the stationary phase
  • Stationary phase does not react with reducing sugars
  • Can be used at elevated temperatures (≤ 80°C) to prevent peak splitting of carbohydrates
  • High efficiency, 3 micron columns offer shorter analysis time and enhanced sensitivity for LC-MS applications
  • Shipped in 85% acetonitrile, 15% water
TSKgel is a registered trademark of Tosoh Corporation
Specifications
Diameter (Metric)1 mm
Length (Metric)10 cm
Max. Pressure60 bar
Particle Size5 μm
pH2 to 7.5
Pore Size100 Å
Solvents0-100% Organic Solvent
Surface Area450 m2/g
Temperature10°C to 80°C
USP TypeL68
Stationary PhaseCarbamoyl
Product LineTSKgel
Shipping ConditionShipped in 85% Acetonitrile, 15% Water
MatrixCarbamoyl Phase/Silica Particle Platform
MaterialStainless Steel Hardware
TechniqueHILIC
Unit SizeEA