Corrosion Resistant Drench Shower, All Stainless Steel and PVC
Corrosion Resistant Drench Shower, All Stainless Steel and PVC
Actual product may vary
Corrosion Resistant Drench Shower, All Stainless Steel and PVC
Haws™ Corrosion Resistant Drench Shower, All Stainless Steel and PVC is an ideal addition to potentially unsafe environments where the body may be exposed to corrosive material
Have Questions?
Catalog NumberMounting
8130Wall
Catalog number 8130
Price (MYR)
10,092.00
EA
Add to cart
Mounting:
Wall
Price (MYR)
10,092.00
EA
Add to cart
  • AXION™ MSR ABS plastic drench showerhead with flow control delivers dispersed flow for uniform drenching
  • Requires minimal maintenance
  • Equipped with test card to record weekly testing
  • Reduced installation time due to pre-built valve assembly
  • Design to mount horizontally or vertically from any wall or ceiling
  • Corrosion resistant Schedule 80 PVC pipe and stainless steel valve makes for a durable long lasting product
Specifications
Certifications/ComplianceANSI Z358.1
Flow Rate20 gpm
Inlet Connection (English)1 in.
Inlet Connection (Metric)2.54 cm
Length (English)22 in.
Length (Metric)55.88 cm
MaterialStainless Steel, PVC
MountingWall
Unit SizeEA