Check Order Status
AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance
Actual product may vary
AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance
Optimized for a variety of applications requiring larger per well volume processing.
Have Questions?
Catalog Number
Pore Size
Volume (Metric)
8075
3.0/0.2 μm
350 μL
Catalog number
8075
Price (MYR)
628.00
CS
Add to cart
Pore Size:
3.0/0.2 μm
Volume (Metric):
350 μL
Price (MYR)
628.00
CS
Add to cart
Plates are constructed of chemically resistant, biologically-inert polypropylene
This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Material (Membrane)
Borosilicate Glass w/o Binder, Supor (PES)
Pore Size
3.0/0.2 μm
Sterility
Non-sterile
Housing Material
Polypropylene Housing
Quantity
10/Pk.
Volume (Metric)
350 μL