PRP-X400
PRP-X400
Actual product may vary
PRP-X400
Have Questions?
Catalog NumberLength (Metric)
79717150 mm
Catalog number 79717
Price (MYR)
-
Length (Metric):
150 mm
Specifications
Diameter (Metric)4.1 mm
Length (Metric)150 mm
Max. Pressure5000 psi
Mobile Phase CompatibilitypH 1-13. 0-100% Aqueous, Organic Modifier
Particle Size7 μm
Quantity1 Ea.
Temperature5°C to 60°C
Stationary PhasePRP-X400
Volume (Metric)2.5 meq
Packing MaterialPSDVB/Sulfonic Acid
MaterialStainless Steel
TypeCation Exchange Column
Unit SizeEach