Acclaim™ C30 HPLC Columns
Acclaim™ C30 HPLC Columns
Actual product may vary
Acclaim™ C30 HPLC Columns
Use for superior separation of hydrophobic structural isomers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
78664250 mm2.1 mm3 μm
075718250 mm4.6 mm5 μm
75718
also known as RP75718
250 mm4.6 mm5 μm
075719150 mm4.6 mm5 μm
75719
also known as RP75719
150 mm4.6 mm5 μm
07572010 mm4.6 mm5 μm
75720
also known as RP75720
10 mm4.6 mm5 μm
075723150 mm4.6 mm3 μm
75723150 mm4.6 mm3 μm
75724
also known as RP75724
150 mm3 mm3 μm
75725
also known as RP75725
150 mm2.1 mm3 μm
075726250 mm3 mm3 μm
75726250 mm3 mm3 μm
Catalog number 78664
Price (MYR)
-
Length (Metric):
250 mm
Diameter (Metric):
2.1 mm
Particle Size:
3 μm
For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim™ C30 LC reversed-phase columns provide superior results. The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.
High Shape Selectivity for the Separation of Structurally Related Isomers
  • Unique selectivity complementary to other reversed-phase columns
  • High efficiency and rugged packing
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8
  • Available in 3 and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 200Å
About the Acclaim Family of HPLC Columns
Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.3 mL/min
Length (Metric)250 mm
Max. Pressure10000 psi (690 bar)
Particle Size3 μm
pH2 to 8
Pore Size200 Å
QuantityEach
Surface Area200 m²/g
Temperature60°C
USP TypeL62
Stationary PhaseC30
Product LineAcclaim
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load13%
Column TypeReversed Phase
EndcappedProprietary
PhaseReversed Phase
Green FeaturesResponsibly packaged
Unit SizeEach