Aquasil™ 3μm C18 HPLC Columns
Aquasil™ 3μm C18 HPLC Columns
Actual product may vary
Aquasil™ 3μm C18 HPLC Columns
Available in limited dimensions. Please contact Customer Service for more details.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
77503-153030150 mm3 mm
77503-104630100 mm4.6 mm
Catalog number 77503-153030
Price (MYR)
-
Length (Metric):
150 mm
Diameter (Metric):
3 mm
Thermo ScientificTM AquasilTM C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
  • Polar endcapped C18 phase
  • Alternative selectively compared to conventional C18 phases
  • Compatible with 100% aqueous mobile phases
  • Ordering Information:
    Other column dimensions are also available. Please call your Customer Service Representative for more information.
    Specifications
    Column FormatAnalytical Column
    Diameter (Metric)3 mm
    For Use WithLC/MS
    Length (Metric)150 mm
    Particle Size3 μm
    pH2 to 8
    Pore Size100 Å
    Surface Area310 m²/g
    USP TypeL1
    Stationary PhaseC18
    Packing MaterialSilica
    Carbon Load12%
    Column TypeReversed Phase
    EndcappedYes
    PhaseReversed Phase
    Unit SizeEach