Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Actual product may vary
Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Equipment)
7600325224 x 0.5/0.6 mL Microliter Tubes24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
7600325024 x 0.2 mL PCR Tubes24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
7600325124 x 0.25/0.4 mL Microliter Tubes24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Catalog number 76003252
Price (MYR)
-
Capacity:
24 x 0.5/0.6 mL Microliter Tubes
For Use With (Equipment):
24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor. Select from a number of Microliter Tube Adapters and PCR adapters for the Thermo Scientific™ 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeMicrotube Adapter
Tube TypeMicrotube
Capacity24 x 0.5/0.6 mL Microliter Tubes
For Use With (Equipment)24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Max. RCF21100
Max. Speed14800
Tube Size0.6/0.5 mL
Unit SizeEA