Sorvall™ Stratos Centrifuge Aerosol-tight Lid for Round Bucket 75008172
Sorvall™ Stratos Centrifuge Aerosol-tight Lid for Round Bucket 75008172
Actual product may vary
Sorvall™ Stratos Centrifuge Aerosol-tight Lid for Round Bucket 75008172
Have Questions?
Catalog NumberFor Use With (Equipment)
75008173Round swinging bucket
Catalog number 75008173
Price (MYR)
-
For Use With (Equipment):
Round swinging bucket
For use with round bucket 75008172 in Four-Place Swinging Bucket Rotor 75003047. Order correct quantity for rotor.
Specifications
For Use With (Equipment)Round swinging bucket
Unit SizeEA