Check Order Status
1000 mL Polypropylene Bottle
1000 mL Polypropylene Bottle
Maximize throughput by combining 1000mL polypropylene bottles with Thermo Scientific rotors.
Have Questions?
Catalog Number
Capacity (Metric)
75007300
1000 mL
Catalog number
75007300
Price (MYR)
-
Capacity (Metric):
1000 mL
Combine 1000mL polypropylene bottles with Thermo Scientific TX-1000 or BIOFlex HC rotors for a complete system and maximize your lab's throughput.
Specifications
Capacity (Metric)
1000 mL
Capacity (English)
33.81 oz.
Material
Polypropylene
Product Type
Centrifuge Bottle
For Use With (Equipment)
TX-1000, BIOFlex HC rotors
Closure Material
Polypropylene