Sorvall LYNX 6000 Superspeed Centrifuge
Sorvall LYNX 6000 Superspeed Centrifuge
Actual product may vary
Sorvall LYNX 6000 Superspeed Centrifuge
The Sorvall LYNX 6000 superspeed centrifuge provides exceptional performance meeting evolving application needs, from academic research to production facilities, with up to 100,605 x g, and up to 6L capacity.
Have Questions?
Catalog NumberElectrical RequirementsDescription
75006590200/240 V 50/60 Hz 30 A, Single PhaseSorvall LYNX 6000 Superspeed Centrifuge
Catalog number 75006590
Price (MYR)
270,996.00
EA
Add to cart
Electrical Requirements:
200/240 V 50/60 Hz 30 A, Single Phase
Description:
Sorvall LYNX 6000 Superspeed Centrifuge
Price (MYR)
270,996.00
EA
Add to cart
The Thermo Scientific™ Sorvall™ LYNX 6000 superspeed centrifuge features powerful technologies with breakthrough simplicity, from push-button security and application flexibility of Thermo Scientific™ Auto-Lock™ rotor exchange, to immediate rotor detection and programming of Thermo Scientific™ Auto-ID instant rotor identification, and the improved ergonomics and performance of Thermo Scientific™ Fiberlite™ carbon fiber rotors. Achieve high-throughput sample processing up to 6L capacity and up to 100,605 x g, with the flexibility to support multiple users and the evolving research needs of a shared laboratory setting, and designed for reliability for consistent results and maximum uptime.
From academic research to bioproduction, support your high performance separations and evolving application needs:
  • Exceptional performance up to 100,605 x g and up to 6L capacity
  • Shortened run set-up time and increased rotor security with rotor innovations
    • Quickly and securely change rotors and applications in as little as 3 seconds with push-button Auto-Lock rotor exchange
    • Auto-ID instant rotor identification automatically loads rotor specifications into centrifuge parameters
  • Choose from 19 optional rotors to find the ideal fit for your application, supporting labware from microplates, to microtubes, to 1L bottles:
    • Lightweight Fiberlite carbon fiber rotors with corrosion- and fatigue-resistance secures the rotor's structural integrity, backed by a 15-year warranty
    • Swinging bucket rotors for versatility and expanded capacity at high RCF with dozens of adapters for research flexibility
    • Complete fixed angle rotor portfolio for commonly used 50mL, 50mL conical, 250mL, 500mL and 1L tubes and bottles with specialty rotor options to address specific protocol requirements
    • Continuous flow/zonal rotors providing scale-up flexibility from superspeed applications to harvesting and clarification
  • Centri-Touch touchscreen interface with bright, highly visible, and durable display simplifies run set-up while providing on-board training and access controls, such as user login with password protection
  • Download the Centri-Vue application for real-time run monitoring and control of 1 or up to 100 centrifuges from your mobile device
  • Advanced technology for resource efficiency including energy use options and Smart Vacuum reduces the power needed for a typical run
  • Expanded levels of biosafety with an optional HEPA filter
  • Compact, non-bolt down design, automatic door opening, and an integrated surface work space optimize everyday centrifuge use
  • Compliant with global standards and designed to assist with GMP/GLP data management
  • Compatible with Centri-Log Plus data management software for FDA 21 CFR Part 11 compliance
Specifications
Depth (English) Exterior31.7 in.
Depth (Metric) Exterior805 mm
Frequency50/60 Hz
Profile (Acceleration/Braking)9 Accel, 10 Decel
TypeSuperspeed Centrifuge
Voltage200/240 V
Capacity6 x 1000 mL
Max. Speed29,000 rpm
Max. RCF100,605 x g
RefrigeratedYes
Height (English) Exterior36.6 in.
Height (Metric) Exterior930 mm
Length (English) Exterior31.7 in.
Length (Metric) Exterior805 mm
Noise Level<59 dBA
Width (English) Exterior27.6 in.
Width (Metric) Exterior700 mm
DescriptionSorvall LYNX 6000 Superspeed Centrifuge
Drive SystemHigh Torque Brushless
Green TechnologyGreen Mode
Other FunctionsMultilingual Selection, Rotor Calculator, On-Board Training Videos, User Logging, User Lock-Out, Automatic Door Opening, Rotor Speed Handle
Run Time99 hours, hold
WarrantyUnit: 1 yr., Drive: 3 yrs., Refrigeration: 5 yrs
Electrical Requirements200/240 V 50/60 Hz 30 A, Single Phase
Certifications/CompliancecULus, CE, ROHS, WEEE
Controller TypeTouchscreen Interface
Dimensions (HWD)36.6 x 27.6 x 31.7 in. (93.0 x 70.0 x 80.5 cm)
Net Weight644 lb. (293 kg)
Program Storage120 programs with alpha-numeric naming
StandardsIEC 61010-2-020 2nd Ed. or later
Temperature Range-20°C to +40°C
Product LineSorvall
Rotor IdentificationAuto-ID Instant
Green FeaturesEnergy efficient, less waste
Unit SizeEA