LH-4000 Fixed Angle Rotor
LH-4000 Fixed Angle Rotor
Actual product may vary
LH-4000 Fixed Angle Rotor
Work within the parameters of existing protocols with this centrifuge rotor, supporting sample processing applications with capacity ranging from 1.5 to 250mL.
Have Questions?
Catalog NumberCapacity
75006475
also known as R75006475
4 x 1000 mL
Catalog number 75006475
also known as R75006475
Price (MYR)
35,160.00
EA
Add to cart
Capacity:
4 x 1000 mL
Price (MYR)
35,160.00
EA
Add to cart
Work within the parameters of existing protocols with the Thermo Scientific™ LH-4000 Fixed Angle Rotor, supporting a diverse array of sample processing applications. Choose from round or DoubleSpin buckets with a wide array of available adapters providing capacity flexibility ranging from 1.5 to 250mL.
Ordering Information:
Order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also ordered separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific Cryofuge 5500i centrifuges.
Note:
Only for use in 230V version
Specifications
DescriptionLH-4000 Fixed Angle Rotor
MaterialStainless Steel Rotor Body, Aluminum Bucket
TypeSwinging Bucket Rotor
Capacity4 x 1000 mL
Max. RCF5,346 x g
Max. Speed4,400 rpm
Radius (Metric)215 mm
Radius (Metric) Min.88 mm
For Use With (Equipment)Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Angle90°
Certifications/ComplianceRound buckets (75006477) and DoubleSpin bucket (75006478) are Biocontainment Certified by CAMR, HPA, Porton Down, U.K.
Min. RCF2,487 x g
BiocontainmentYes
Unit SizeEA