Polypropylene Centrifuge Bottles
Polypropylene Centrifuge Bottles
Actual product may vary
Polypropylene Centrifuge Bottles
Polypropylene bottle with cap
Have Questions?
Catalog NumberCapacity (Metric)
75006443750 mL
Catalog number 75006443
Price (MYR)
-
Capacity (Metric):
750 mL
Polypropylene bottle with cap
Specifications
Capacity (Metric)750 mL
Capacity (English)25.36 oz.
ColorClear
MaterialPolypropylene
Closure ColorBlue
ShapeRound
Product TypeCentrifuge Bottle
Closure MaterialPolypropylene
Closure TypeScrew Cap
Unit SizeEA