Check Order Status
1.5/2 mL Microtube Adapter for MicroClick 18 x 5 Rotor
1.5/2 mL Microtube Adapter for MicroClick 18 x 5 Rotor
Accommodate 1.5/2mL microtubes in the Thermo Scientific™ MicroClick 18 x 5 rotor.
Have Questions?
Catalog Number
Capacity
75005756
18 x 1.5/2 mL
Catalog number
75005756
HSN
39269099
Price (MYR)
0.00
EA
Add to cart
Capacity:
18 x 1.5/2 mL
Price (MYR)
0.00
EA
Add to cart
1.5/2mL adapters for use with the Thermo Scientific MicroClick 18 x 5 rotor.
Specifications
Type
Microtube Adapter
Tube Type
Microtube
Capacity
18 x 1.5/2 mL
Max. Tube Diameter
45 mm
Max. Tube Length
11 mm
For Use With (Equipment)
MicroClick 18 x 5 rotor
Description
1.5/2mL Microtube Adapter
Tube Size
1.5/2 mL