145 mL Bottle for TX-150 Rotor
145 mL Bottle for TX-150 Rotor
Actual product may vary
145 mL Bottle for TX-150 Rotor
Bottle directly fits into the TX-150 rotor
Have Questions?
Catalog NumberCapacity (Metric)
75005734125 mL
Catalog number 75005734
Price (MYR)
-
Capacity (Metric):
125 mL
Bottle directly fits into the TX-150 rotor
Specifications
Capacity (Metric)125 mL
Description125 mL bottle directly fit into TX-150 rotor
MaterialPolypropylene
No. per Pack4
AutoclavableAutoclavable
Height (Metric)99 mm
Product TypeCentrifuge Bottle
For Use With (Equipment)TX-150 Rotor
Height (English)3.89 in.
Closure MaterialPolypropylene
Diameter (Metric)50 mm
Unit SizeEA