Check Order Status
Buckets and Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Actual product may vary
Buckets and Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Round and conical buckets offered in sets of four
Have Questions?
Change view
Catalog Number
Type
Capacity
75005707
Bucket Lid
Not Applicable
75005702
Bucket
4 x 145 mL
75005703
Bucket
8 x 50 mL
3 Options
Catalog number
75005707
Price (MYR)
-
Type:
Bucket Lid
Capacity:
Not Applicable
Round and conical buckets offered in sets of four
Specifications
For Use With (Equipment)
Round Buckets
Capacity
Not Applicable
Type
Bucket Lid
Quantity
Set of 4
Description
ClickSeal Biocontainment Lids for Round Buckets