M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Actual product may vary
M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Spin up to 4 microplates or 2 deepwell plates with the convenience of transportable microplate carriers.
Have Questions?
Catalog NumberCapacity
750057064 Standard or 2 Deepwell Microplates
Catalog number 75005706
Price (MYR)
-
Capacity:
4 Standard or 2 Deepwell Microplates
Ideally suited for quick spins—centrifuge 4 microplates or 2 midi-deepwell plates with the convenience of transportable carriers using the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Spin up to 4 standard or 2 midi-deepwell microplates with optional ClickSeal biocontainment lids, certified by Public Health England, Porton Down, UK.
Non-shielded design provides quick access to microplates.
  • Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers:
  • Trouble-free rotor installation and removal
    • No tools are required
    • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Unsealed buckets (75005723) and sealed buckets (75005721) are sold separately. Contact your sales representative for more information.
Specifications
DescriptionM10 Microplate Swinging Bucket Rotor
TypeSwinging Bucket Rotor
Capacity4 Standard or 2 Deepwell Microplates
Max. RCF2,576 x g
Max. Speed4,400 rpm
Radius (Metric)119 mm
K Factor13103
For Use With (Equipment)Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle90°
BiocontainmentYes
Unit SizeEA