Sorvall™ RC4 General Purpose Floor Model Centrifuge
Sorvall™ RC4 General Purpose Floor Model Centrifuge
Actual product may vary
Sorvall™ RC4 General Purpose Floor Model Centrifuge
Process the optimal capacity for small to medium size blood processing centers with the ability to spin 4 or 8 blood bags in a wide range of bag configurations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical Requirements
75004481200/230 V 50/60 Hz
75004473230 V 50 Hz
Catalog number 75004481
Price (MYR)
139,095.00
EA
Add to cart
Electrical Requirements:
200/230 V 50/60 Hz
Price (MYR)
139,095.00
EA
Add to cart
Process the optimal capacity for small to medium size blood processing centers with the ability to spin 4 or 8 blood bags in a wide range of bag configurations when using the Thermo Scientific™ Sorvall™ RC4 Low-Speed Floor Model Centrifuge, a flexible refrigerated centrifuge for blood banking and clinical applications. Using flexible accessory systems, it spins everything from doubles to 500mL quints with in-line leukoreduction filters. A versatile performer, the RC-4 is in a class of its own.
High-Throughput and Versatiliy
  • Spins larger numbers of tubes at higher RCFs than competing models
  • Powerful induction drive and optional swinging bucket rotor (which accommodates DoubleSpin buckets) combine to provide higher RCFs and capacities for the most popular sample formats
  • Exceptional capacity for all popular sample formats, including 15 and 50mL tissue culture tubes, and many others. A wide range of rotors makes it suitable for biochemistry, biotechnology, microbiology, pharmaceutical and blood processing labs
Safe Operation
  • The RC4 meets stringent international safety standards. It offers a wide selection of independently tested biocontainment rotors that comply with rigorous international guidelines for aerosol containment of bacteria and viruses
Optimal Run Stability
  • The Thermo Scientific™ SMARTspin™ Imbalance Detection System automatically brakes unit if an excessive imbalance is detected, for example, due to tube leakage
  • The reliable imbalance detection means that even large swinging-bucket rotors can operate safely with a 30g imbalance
Easy to Use
  • EASYcontrol™ II Soft-Touch Keypad is located at the front of the instrument, eliminating awkward reaching
  • Soft-Touch Lid Lock allows effortless opening and closing
  • Intuitive operator keypad allows you to recall up to 9 protocols
Key Features
  • Max capacity of 4 x 1000mL bottles, 8 x 500mL blood bags or 4 to 8 standard microplates
  • Fastest processing and highest throughput of any 4L low-speed centrifuge
  • Unmatched RCFs with fixed-angle rotors for bacterial, tissue culture and general pelleting needs
  • Enhanced user safety: choose from 4 biocontainment rotors accommodating samples from 1.5 to 1000mL
  • HIGHplate™ rotor spins DNA/RNA filter plates to 85mm in height
  • Optimum run stability with SMARTspin™ Imbalance Detection System
  • Reduced maintenance with brushless induction drive
  • Whisper quiet operation at a noise level under 58dBA
    • Compatible with Centri-Log Plus data management software for FDA 21 CFR Part 11 compliance
Requires:
Rotor (sold separately)
Specifications
Controller TypeMicroprocessor via EASYcontrol II
Dimensions (HWD)37.8 x 26.8 x 28.3 in. (96 x 68 x 72 cm)
Frequency50/60 Hz
Net Weight631 lb. (286 kg)
Temperature Range-9°C to +40°C
TypeCentrifuge
Capacity4 x 1000 mL or 8 x 500 mL Blood Bags or 4 x 8 Microplates
Product LineSorvall
RefrigeratedCFC free
Max. Speed10,000 rpm
Max. RCF15,320 x g
Amperage16 A
Height (English) Exterior37.8 in.
Height (Metric) Exterior96 cm
Imbalance DetectorOptimum run stability with SMARTspin Imbalance Detection System
Length (English) Exterior28.3 in.
Length (Metric) Exterior72 cm
Run Time0 to 99 min., plus HOLD
Width (English) Exterior26.8 in.
Width (Metric) Exterior68 cm
Certifications/ComplianceDesigned and tested in accordance with EN 61 010-1, EN 61 010-2-020, EN 50 081-1, EN 50-082-1. CSA
DescriptionSorvall RC4 General Purpose Floor Model Centrifuge
Electrical Requirements200/230 V 50/60 Hz
Profile (Acceleration/Braking)9/9
Unit SizeEA