2000mL BIOS Swinging Bucket Rotors
2000mL BIOS Swinging Bucket Rotors
Actual product may vary
2000mL BIOS Swinging Bucket Rotors
2000mL BIOS Swinging Bucket Rotors
2000mL BIOS Swinging Bucket Rotors
The unique 8 x 2000mL (16L) capacity of the Thermo Scientific™ 2000mL BIOS swinging bucket rotors enhance the productivity of your bioprocessing methods.
Have Questions?
Catalog NumberCapacityIncludes
750039828 x 2000 mLRotor, Oval Buckets, 2000 mL Bottles
Catalog number 75003982
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
8 x 2000 mL
Includes:
Rotor, Oval Buckets, 2000 mL Bottles
Thermo Scientific™ 2000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. The rotor, buckets and bottles which are provided as a set, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:
  • 6 x 2000mL (12L) capacity at 4,700 max rpm (7,187 x g max RCF) operation;
  • 8 x 2000mL (16L)  capacity at 3,900 max rpm (5,374 x g max RCF) operation
  • Easy-to-use innovative 2000mL bucket and bottle with a wide mouth design requires no bridge and are resistant to 1000 cycles
  • Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 60% when compared with non-windshielded rotors
  • Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
  • 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
MaterialStainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle
TypeSwinging Bucket Rotor
Capacity8 x 2000 mL
Max. RCF5,374 x g
Max. Speed3,900 rpm
For Use With (Equipment)Thermo Scientific Sorvall BIOS 16 Centrifuge
IncludesRotor, Oval Buckets, 2000 mL Bottles
Unit SizeEA