HAEMAFlex™ 12/16 Swinging Bucket Rotors
HAEMAFlex™ 12/16 Swinging Bucket Rotors
Actual product may vary
HAEMAFlex™ 12/16 Swinging Bucket Rotors
Maximize total blood bags per spin with the unique 16 x 500mL blood bag capacity of the Thermo Scientific™ HAEMAFlex™ 12/16 rotors for the new Thermo Scientific™ blood banking centrifuges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
75003882HAEMAFlex 16 Swinging Bucket Rotor
75003862HAEMAFlex 12 Swinging Bucket Rotor
Catalog number 75003882
Price (MYR)
178,017.00
EA
Add to cart
Description:
HAEMAFlex 16 Swinging Bucket Rotor
Price (MYR)
178,017.00
EA
Add to cart
For high-throughput blood processing needs, the Thermo Scientific™ HAEMAFlex™ 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the Thermo Scientific™ HAEMAFlex™ 16 rotor and the same bucket selections.
The rotors HAEMFlex 12 and 16 can be equipped with adapters to spin the most common blood collection tubes, conical tubes and plates, using oval buckets (750003964).
High-throughput sample processing with the flexibility to match your current and future needs with 12 or 16 x 500mL blood bag (12 or 16 x 1000mL) capacity, utilizing the same bucket system.
We also offer a range of adapters allowing the centrifugation of most common tubes and microplates, using the same rotor body and oval buckets (75003964).
These windshielded rotors feature Thermo Scientific™ Eco-Spin™ technology, providing energy savings up to 60%, based on an engineering evaluation of windshielded and non-windshielded designs of the same rotor body.
Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ functions enabling instant rotor lid opening, storage and rotor identification to save time and protects the integrity of your samples.
Rotor buckets with Thermo Scientific™ Dura-Coat™ nickel-coating are also available, providing protection against corrosion due to moisture, chemicals or alkaline solutions that otherwise can weaken the structural integrity of a metal rotor bucket.
Specifications
DescriptionHAEMAFlex 16 Swinging Bucket Rotor
MaterialStainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle
TypeSwinging Bucket Rotor
Capacity16 x 500 mL blood bags (16 x 1000 mL), 416 x 5/7 mL blood tubes (oval buckets 75003964 required)
Max. RCF5,374 x g
Max. Speed3,900 rpm
For Use With (Equipment)Thermo Scientific Cryofuge 8 and 16 and Sorvall BP 8 and 16 Centrifuges
Unit SizeEA