TX-750 Round Bucket Adapters
TX-750 Round Bucket Adapters
Actual product may vary
TX-750 Round Bucket Adapters
TX-750 Round Bucket Adapters
TX-750 Round Bucket Adapters
TX-750 Round Bucket Adapters
TX-750 Round Bucket Adapters
For enhanced functionality of TX-750 Rotor
Have Questions?
Catalog NumberCapacity
7500382450 mL Conical or Skirted Tubes
Catalog number 75003824
Price (MYR)
-
Capacity:
50 mL Conical or Skirted Tubes
For enhanced functionality of TX-750 Rotor, these adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
Tube TypeConical, Skirted
Capacity50 mL Conical or Skirted Tubes
Max. Tube Diameter30 mm
Max. Tube Length120 mm
For Use With (Equipment)TX-750 Rotor
Vessels per Adapter20 x 50 mL
Unit SizeEA