BIOShield™ 720 Rotor Tube Adapters
BIOShield™ 720 Rotor Tube Adapters
Actual product may vary
BIOShield™ 720 Rotor Tube Adapters
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
750038215/7 mL Blood Collection Tube
7500367750 mL Conical Tube
Catalog number 75003821
Price (MYR)
-
Capacity:
5/7 mL Blood Collection Tube
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
Specifications
TypeRotor Tube Adapter
Tube Type5/7 mL Blood Collection Tube
Capacity5/7 mL Blood Collection Tube
Unit SizeEA